ENA053246827 Enamine

8-chloro-2,3-dihydro-1,4-benzodioxine-6-carbonitrile

  • MDL number MFCD06655478
  • Cas Number : 852389-10-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-156
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12756

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12756
Availability details for
Quantity
  • Availability
  • Add to Cart