ENA053246830 Enamine

3-{4-oxo-1-phenyl-1H,4H,5H-pyrazolo[3,4-d]pyrimidin-6-yl}propanoic acid

  • MDL number MFCD01918224
  • Cas Number : 307341-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-141
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12759

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12759
Availability details for
Quantity
  • Availability
  • Add to Cart