ENA053246835 Enamine

6-methyl-7H,8H-[1,2,4]triazolo[4,3-b][1,2,4]triazin-7-one

  • MDL number MFCD06655483
  • Cas Number : 19542-10-0
  • Molecular Weight : 151.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195-196
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12764

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12764
Availability details for
Quantity
  • Availability
  • Add to Cart