ENA053246841 Enamine

2-chloro-N-(prop-2-en-1-yl)-N-[(thiophen-2-yl)methyl]acetamide

  • MDL number MFCD06655488
  • Cas Number : 852399-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-12770

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12770
Availability details for
Quantity
  • Availability
  • Add to Cart