ENA053246845 Enamine

5-(piperidin-1-ylsulfonyl)pyridine-2-thiol

  • MDL number MFCD06655491
  • Cas Number : 852400-11-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218...221
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12774

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12774
Availability details for
Quantity
  • Availability
  • Add to Cart