ENA053246846 Enamine

ethyl 2-amino-6,6-dioxo-4H,5H,7H-6lambda6-thieno[2,3-c]thiopyran-3-carboxylate

  • MDL number MFCD06655492
  • Cas Number : 474843-59-9
  • Molecular Weight : 275.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185-186
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-12775

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12775
Availability details for
Quantity
  • Availability
  • Add to Cart