ENA053246847 Enamine

7-methyl-5,6,7,8-tetrahydro[1]benzothieno[2,3-d][1,2,3]triazin-4(3H)-one

  • MDL number MFCD03621856
  • Cas Number : 496038-39-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12776

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12776
Availability details for
Quantity
  • Availability
  • Add to Cart