ENA053246848 Enamine

3-(3-methylphenyl)-2-thien-2-ylacrylic acid

  • MDL number MFCD06655493
  • Cas Number : 852400-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158...160
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12777

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12777
Availability details for
Quantity
  • Availability
  • Add to Cart