ENA053246849 Enamine

2-({5,6-dimethylthieno[2,3-d]pyrimidin-4-yl}amino)propanoic acid

  • MDL number MFCD06255213
  • Cas Number : 955966-34-4
  • Molecular Weight : 251.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12778

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12778
Availability details for
Quantity
  • Availability
  • Add to Cart