ENA053246852 Enamine

1-(1,3-benzothiazol-2-yl)ethan-1-one

  • MDL number MFCD00192994
  • Cas Number : 1629-78-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107-110
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-12781

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12781
Availability details for
Quantity
  • Availability
  • Add to Cart