ENA053246854 Enamine

2-(chloromethyl)-4-[(2-chlorophenyl)methyl]morpholine

  • MDL number MFCD06655496
  • Cas Number : 852400-07-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-12783

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12783
Availability details for
Quantity
  • Availability
  • Add to Cart