ENA053246860 Enamine

1-[(1H-1,3-benzodiazol-2-yl)methyl]cyclopentane-1-carboxylic acid

  • MDL number MFCD06655500
  • Cas Number : 852399-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12789

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12789
Availability details for
Quantity
  • Availability
  • Add to Cart