ENA053246866 Enamine

2-[5-(4-fluorophenyl)-2-oxo-2,3-dihydro-1,3,4-oxadiazol-3-yl]acetic acid

  • MDL number MFCD06655503
  • Cas Number : 852389-17-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-111
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-12795

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12795
Availability details for
Quantity
  • Availability
  • Add to Cart