ENA053246867 Enamine

(8-chloro-2,3-dihydro-1,4-benzodioxin-6-yl)methanol

  • MDL number MFCD06655504
  • Cas Number : 852399-55-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62-63
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12796

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12796
Availability details for
Quantity
  • Availability
  • Add to Cart