ENA053246869 Enamine

6-methyl-1-(2-nitrophenyl)-4-oxo-1,4-dihydropyridazine-3-carboxylic acid

  • MDL number MFCD06655506
  • Cas Number : 154777-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12798

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12798
Availability details for
Quantity
  • Availability
  • Add to Cart