ENA053246877 Enamine

2-chloro-N-(1,1-dioxo-1lambda6-thiolan-3-yl)-N-[(oxolan-2-yl)methyl]acetamide

  • MDL number MFCD05668165
  • Cas Number : 852399-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-12806

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12806
Availability details for
Quantity
  • Availability
  • Add to Cart