ENA053246898 Enamine

5-[(2-chlorophenyl)amino]-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD00178429
  • Cas Number : 37844-22-7
  • Molecular Weight : 243.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174-175
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12827

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12827
Availability details for
Quantity
  • Availability
  • Add to Cart