ENA053246899 Enamine

5-[(3-fluorophenyl)amino]-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD06655521
  • Cas Number : 852400-49-8
  • Molecular Weight : 227.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206-207
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12828

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12828
Availability details for
Quantity
  • Availability
  • Add to Cart