ENA053246901 Enamine

5-{[2-(trifluoromethyl)phenyl]amino}-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD06655523
  • Cas Number : 852399-74-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172-173
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12830

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12830
Availability details for
Quantity
  • Availability
  • Add to Cart