ENA053246903 Enamine

5-[(3-chlorophenyl)amino]-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD06655525
  • Cas Number : 76462-59-4
  • Molecular Weight : 243.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210-211
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12832

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12832
Availability details for
Quantity
  • Availability
  • Add to Cart