ENA053246904 Enamine

3-(2,4-dimethylphenyl)-2-hydrazinoquinazolin-4(3H)-one

  • MDL number MFCD06655526
  • Cas Number : 77066-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12833

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12833
Availability details for
Quantity
  • Availability
  • Add to Cart