ENA053246908 Enamine

3-(3-ethoxypropyl)-2-hydrazinoquinazolin-4(3H)-one

  • MDL number MFCD06655530
  • Cas Number : 852399-86-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12837

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12837
Availability details for
Quantity
  • Availability
  • Add to Cart