ENA053246917 Enamine

N-phenyl-1,3-benzothiazol-2-amine

  • MDL number MFCD00453249
  • Cas Number : 1843-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139...141
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12846

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12846
Availability details for
Quantity
  • Availability
  • Add to Cart