ENA053246919 Enamine

N-(4-methylphenyl)-1,3-benzothiazol-2-amine

  • MDL number MFCD01830676
  • Cas Number : 70785-26-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-12848

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12848
Availability details for
Quantity
  • Availability
  • Add to Cart