ENA053246922 Enamine

N-(2,4-dimethylphenyl)-1,3-benzothiazol-2-amine

  • MDL number MFCD00115419
  • Cas Number : 216502-42-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-12851

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12851
Availability details for
Quantity
  • Availability
  • Add to Cart