ENA053246925 Enamine

N-1,3-benzothiazol-2-yl-2-chloro-N-(4-methylphenyl)acetamide

  • MDL number MFCD06655536
  • Cas Number : 852399-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12854

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12854
Availability details for
Quantity
  • Availability
  • Add to Cart