ENA053246931 Enamine

5-(4-methylphenyl)-1-phenyl-1H-imidazole-2-thiol

  • MDL number MFCD06655540
  • Cas Number : 852400-51-2
  • Molecular Weight : 266.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12860

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12860
Availability details for
Quantity
  • Availability
  • Add to Cart