ENA053246942 Enamine

2-chloro-N-[3-cyano-1-(3-fluorophenyl)-4,5-dimethyl-1H-pyrrol-2-yl]acetamide

  • MDL number MFCD06655544
  • Cas Number : 852399-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-12871

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12871
Availability details for
Quantity
  • Availability
  • Add to Cart