ENA053246943 Enamine

2-chloro-N-(3-cyano-1-cyclohexyl-4,5-dimethyl-1H-pyrrol-2-yl)acetamide

  • MDL number MFCD06655545
  • Cas Number : 852399-64-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-12872

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12872
Availability details for
Quantity
  • Availability
  • Add to Cart