ENA053246947 Enamine

N-(3,4-dimethylphenyl)-1,3-benzothiazol-2-amine

  • MDL number MFCD06655549
  • Cas Number : 852399-77-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-12876

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12876
Availability details for
Quantity
  • Availability
  • Add to Cart