ENA053246950 Enamine

2-(morpholin-4-yl)-5-sulfamoylbenzoic acid

  • MDL number MFCD06655551
  • Cas Number : 65194-68-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220-221
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12879

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12879
Availability details for
Quantity
  • Availability
  • Add to Cart