ENA053246953 Enamine

3-acetyl-7-(diethylamino)-2H-chromen-2-one

  • MDL number MFCD00068687
  • Cas Number : 74696-96-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149-152
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-12882

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12882
Availability details for
Quantity
  • Availability
  • Add to Cart