ENA053246975 Enamine

N'-(chloroacetyl)-4-methylbenzohydrazide

  • MDL number MFCD00445423
  • Cas Number : 199938-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12904

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12904
Availability details for
Quantity
  • Availability
  • Add to Cart