ENA053246976 Enamine

2-{[(3-bromophenyl)methyl]sulfanyl}acetic acid

  • MDL number MFCD00486087
  • Cas Number : 98288-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70-71
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-12905

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12905
Availability details for
Quantity
  • Availability
  • Add to Cart