ENA053246978 Enamine

4-(bromoacetyl)-N,N-dimethylbenzenesulfonamide

  • MDL number MFCD06682723
  • Cas Number : 89102-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90...92
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12907

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12907
Availability details for
Quantity
  • Availability
  • Add to Cart