ENA053246979 Enamine

2-chloro-N-(3-{5,6-dimethyl-4-oxo-3H,4H-thieno[2,3-d]pyrimidin-2-yl}phenyl)acetamide

  • MDL number MFCD06655558
  • Cas Number : 852033-50-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-12908

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12908
Availability details for
Quantity
  • Availability
  • Add to Cart