ENA053246980 Enamine

1-methylpiperidin-4-amine

  • MDL number MFCD01570549
  • Cas Number : 41838-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12909

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-12909
pictograms GHS02,GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart