ENA053246989 Enamine

4-(4-acetylpiperazin-1-yl)-3-nitrobenzoic acid

  • MDL number MFCD02111682
  • Cas Number : 309943-00-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12918

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12918
Availability details for
Quantity
  • Availability
  • Add to Cart