ENA053246990 Enamine

4-acetyl-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid

  • MDL number MFCD06655565
  • Cas Number : 92288-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-183
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12919

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12919
Availability details for
Quantity
  • Availability
  • Add to Cart