ENA053247002 Enamine

[2-(diethylamino)ethyl][(thiophen-2-yl)methyl]amine

  • MDL number MFCD06655572
  • Cas Number : 852706-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-12931

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12931
Availability details for
Quantity
  • Availability
  • Add to Cart