ENA053247005 Enamine

N-(4,5-dimethoxy-2-methylbenzyl)-N-methylamine

  • MDL number MFCD06655573
  • Cas Number : 852706-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180...182
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12934

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12934
Availability details for
Quantity
  • Availability
  • Add to Cart