ENA053247010 Enamine

4-(3-fluoro-4-methoxyphenyl)-1,3-thiazol-2-amine

  • MDL number MFCD02663993
  • Cas Number : 716-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12939

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12939
Availability details for
Quantity
  • Availability
  • Add to Cart