ENA053247012 Enamine

4-phenyl-1H-imidazole-1,2-diamine

  • MDL number MFCD00778400
  • Cas Number : 15970-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 258...260
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12941

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12941
Availability details for
Quantity
  • Availability
  • Add to Cart