ENA053247027 Enamine

2-[2-(1,3-dithiolan-2-yl)phenoxy]acetic acid

  • MDL number MFCD06655770
  • Cas Number : 885524-23-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-13288

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13288
Availability details for
Quantity
  • Availability
  • Add to Cart