ENA053247028 Enamine

methyl 4-isothiocyanatobutanoate

  • MDL number MFCD00060423
  • Cas Number : 62210-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 229...231
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13314

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-13314
pictograms GHS05,GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart