ENA053247030 Enamine

4-ethyl 2-methyl 5-(2-chloroacetamido)-3-(2-methoxy-2-oxoethyl)thiophene-2,4-dicarboxylate

  • MDL number MFCD06655847
  • Cas Number : 885524-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-121
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-13422

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13422
Availability details for
Quantity
  • Availability
  • Add to Cart