ENA053247033 Enamine

5-[(1H-1,2,4-triazol-1-yl)methyl]-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD06655585
  • Cas Number : 852933-13-2
  • Molecular Weight : 183.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-12953

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12953
Availability details for
Quantity
  • Availability
  • Add to Cart