ENA053247034 Enamine

3-(4,4-dimethyl-2,5-dioxoimidazolidin-1-yl)propanohydrazide

  • MDL number MFCD06655586
  • Cas Number : 90345-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176...178
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12954

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12954
Availability details for
Quantity
  • Availability
  • Add to Cart