ENA053247038 Enamine

2-chloro-N-ethyl-N-[(thiophen-2-yl)methyl]acetamide

  • MDL number MFCD06655588
  • Cas Number : 852706-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12958

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12958
Availability details for
Quantity
  • Availability
  • Add to Cart