ENA053247040 Enamine

2-(2,5-dimethyl-1H-pyrrol-1-yl)-4,5-dimethylthiophene-3-carboxylic acid

  • MDL number MFCD03167268
  • Cas Number : 432530-64-8
  • Molecular Weight : 249.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12960

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12960
Availability details for
Quantity
  • Availability
  • Add to Cart